Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human S1P3/EDG-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB75241SP
This item is not returnable.
View return policy
Description
S1P3/EDG-3 Monoclonal specifically detects S1P3/EDG-3 in Human samples. It is validated for Immunohistochemistry.Specifications
S1P3/EDG-3 | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
Q99500 | |
S1PR3 | |
NS0 mouse myeloma cell line transfected with human S1P 3EDG-3 Accession # Q99500 | |
25 μg | |
Primary | |
Detects human S1P 3EDG-3 in direct ELISAs. | |
Human | |
Purified |
Immunohistochemistry | |
776807 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EDG3, EDG-3, EDG3FLJ37523, Endothelial differentiation G-protein coupled receptor 3, endothelial differentiation, sphingolipid G-protein-coupled receptor, 3, FLJ93220, G protein-coupled receptor, endothelial differentiation gene-3, LPB3, MGC71696, S1P receptor 3, S1P receptor Edg-3, S1P receptor EDG3, S1P3, S1PR3, sphingosine 1-phosphate receptor 3, Sphingosine 1-phosphate receptor Edg-3, sphingosine-1-phosphate receptor 3 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1903 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction