Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human S1P4/EDG-6 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB10321V10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB10321V10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB10321V10 Supplier R&D Systems Supplier No. FAB10321V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

S1P4/EDG-6 Monoclonal specifically detects S1P4/EDG-6 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen S1P4/EDG-6
Applications Flow Cytometry
Classification Monoclonal
Clone 1012512
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. O95977
Gene Alias EDG6, EDG6S1P receptor Edg-6, Endothelial differentiation G-protein coupled receptor 6, endothelial differentiation, G-protein-coupled receptor 6, endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor6,endothelial differentiation, G protein coupled receptor 6, S1P receptor 4, S1P4LPC1, S1PR4, SLP4, sphingosine 1-phosphate receptor 4, Sphingosine 1-phosphate receptor Edg-6, sphingosine-1-phosphate receptor 4
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human S1P4/EDG-6, Met1-Ile384, Accession # O95977
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8698
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.