Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human S1P5/EDG-8 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB3964N100UG
This item is not returnable.
View return policy
Description
S1P5/EDG-8 Monoclonal specifically detects S1P5/EDG-8 in Human samples. It is validated for Flow Cytometry.Specifications
S1P5/EDG-8 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9H228 | |
S1PR5 | |
BaF3 mouse pro-B cell line transfected with human S1P 5EDG-8 Met1-Asp398 Accession # Q9H228 | |
RUO | |
53637 | |
Human | |
IgG2b |
Flow Cytometry | |
282503 | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Edg-8, EDG8, EDG8S1P receptor Edg-8, Endothelial differentiation G-protein-coupled receptor 8, endothelial differentiation, sphingolipid G-protein-coupled receptor, 8, LPB4, S1P receptor 5, S1P5, S1PR5, sphingosine 1-phosphate receptor 5, sphingosine 1-phosphate receptor EDG8, Sphingosine 1-phosphate receptor Edg-8, sphingosine-1-phosphate receptor 5, SPPR-1, SPPR-2 | |
Mouse | |
100 μg | |
Primary | |
Detects human S1P 5EDG-8. Stains human S1P 5EDG-8 transfectants but not irrelevant transfectants. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction