Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SACS Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF8014
This item is not returnable.
View return policy
Description
SACS Polyclonal specifically detects SACS in Human samples. It is validated for Immunohistochemistry.Specifications
SACS | |
Polyclonal | |
Unconjugated | |
Q9NZJ4 | |
SACS | |
E. coli-derived recombinant human SACS Asn4402-Val4579 Accession # Q9NZJ4 | |
100 μg | |
Primary | |
Detects human SACS in direct ELISAs. | |
Human | |
IgG |
Immunohistochemistry | |
LYOPH | |
Immunohistochemistry 5-15 ug/mL | |
ARSACS, dnaJ homolog subfamily C member 29, DNAJC29, KIAA0730, Sacsin, spastic ataxia of Charlevoix-Saguenay (sacsin) | |
Sheep | |
Affinity Purified | |
RUO | |
26278 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction