Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SAGE1 Partial ORF (NP_061136, 357 a.a. - 455 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7101064
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

This gene belongs to a class of genes that are activated in tumors. These genes are expressed in tumors of different histologic types but not in normal tissues, except for spermatogenic cells and, for some, placenta. The proteins encoded by these genes appear to be strictly tumor specific, and hence may be excellent sources of antigens for cancer immunotherapy. This gene is expressed in sarcomas. [provided by RefSeq]

Sequence: PLPSNALSTVLPGLAYLATADMPAMSTRDQHATIIHNLREEKKDNSQPTPDNVLSAVTPELINLAGAGIPPMSTRDQYATVNHHVHEARMENGQRKQDN

Specifications

Accession Number NP_061136
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 55511
Molecular Weight (g/mol) 36.63kDa
Name SAGE1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen PLPSNALSTVLPGLAYLATADMPAMSTRDQHATIIHNLREEKKDNSQPTPDNVLSAVTPELINLAGAGIPPMSTRDQYATVNHHVHEARMENGQRKQDN
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias SAGE
Common Name SAGE1
Gene Symbol SAGE1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.