Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SALM2/LRFN1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5669
This item is not returnable.
View return policy
Description
SALM2/LRFN1 Polyclonal specifically detects SALM2/LRFN1 in Human samples. It is validated for Western Blot.Specifications
SALM2/LRFN1 | |
Polyclonal | |
Unconjugated | |
KIAA1484, leucine rich repeat and fibronectin type III domain containing 1, leucine-rich repeat and fibronectin type III domain-containing protein 1, LRFN1, SALM2, synaptic adhesion-like molecule 2 | |
Sheep | |
Affinity Purified | |
RUO | |
57622 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
LRFN1 | |
Chinese hamster ovary cell line CHO-derived recombinant human SALM2/LRFN1 | |
100 μg | |
Primary | |
Detects human SALM2/LRFN1 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) SALM3 and rhSALM4 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction