Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SALM3/LRFN4 Antibody, R&D Systems™

Catalog No. MAB5445
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5445 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5445 Supplier R&D Systems Supplier No. MAB5445
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SALM3/LRFN4 Monoclonal specifically detects SALM3/LRFN4 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen SALM3/LRFN4
Applications Immunohistochemistry
Classification Monoclonal
Clone 578102
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q6PJG9
Gene Alias FIGLER6, LRFN4, SALM3
Gene Symbols LRFN4
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human SALM3/LRFN4 Cys17-Leu518 Accession # Q6PJG9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 78999
Test Specificity Detects human SALM3/LRFN4 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) SALM1, approximately 20-25% cross-reactivity with recombinant human rhSALM2, and no cross-reactivity with rhSALM4 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.