Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SCARA5 Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB4900V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB4900V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB4900V100 Supplier R&D Systems Supplier No. FAB4900V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SCARA5 Monoclonal specifically detects SCARA5 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen SCARA5
Applications Flow Cytometry
Classification Monoclonal
Clone 673527
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q6ZMJ2
Gene Alias FLJ23907, MGC45780, scavenger receptor class A, member 5 (putative), Tesr, testis expressed scavenger receptor, UNQ2938/PRO28700
Gene Symbols SCARA5
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human SCARA5 Arg83-His495 Accession # Q6ZMJ2
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286133
Test Specificity Detects human SCARA5 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse SCARA5 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.