Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SCD-1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7550
This item is not returnable.
View return policy
Description
SCD-1 Polyclonal specifically detects SCD-1 in Human samples. It is validated for Western Blot, Knockout Validated.Specifications
| SCD-1 | |
| Polyclonal | |
| Unconjugated | |
| O00767 | |
| SCD | |
| E. coli-derived recombinant human SCD-1 Ala141-Gly221 Accession # O00767 | |
| 100 μg | |
| Primary | |
| Detects human SCD-1 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human SCD-5 and recombinant mouse SCD-1 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| FADS5, MSTP008, SCD1, SCDOS, Stearoyl-CoA desaturase | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 6319 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction