Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CD137 (4-1BB) (Soluble) ELISA Kit

Catalog No. 5018240
Encompass_Preferred
Click to view available options
Quantity:
10 x 96 Tests
96 Tests

Includes

Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD137
Biotin-Conjugate anti-human sCD137 polyclonal antibody
Streptavidin-HRP
Human sCD137 Standard lyophilized, 100ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

ELISA

The Human Soluble CD137 (4-1BB) (Hu CD137) ELISA quantitates Hu CD137 in human serum or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu CD137. Principle of the method The Human CD137 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

CD137, also known as TNFRSF9 or 4-1BB, is an inducible costimulatory molecule expressed mainly on activated T cells. Its ligand, known as 4-1BBL, is expressed on activated macrophages, mature B cells, hematopoietic stem cells, and myeloid progenitor cells. CD137 signaling leads to maintaining the survival of activated T cells and CD8+ memory T cells, and clonal expansion of T cells, but also to suppressing myelopoiesis and dendritic cell development. Triggered CD137 induces a cytokine release profile regulating peripheral monocyte survival. Soluble forms of CD137 may provide negative control mechanism for some immune responses.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q07011
Assay Range 0.78 to 50 ng/mL
Assay Sensitivity 0.2 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Serum, Supernatant
Target Species Human
For Use With (Application) ELISA
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 3604
Gene Symbol 4-1 BB,CD137,CDw137,ILA
Interassay CV 9.9%
Intraassay CV 7.1
Kit Contents Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Synonym 4-1 BB Ligand Receptor, TNFRSF9, ILA
Target 4-1 BB (CD137)
Test Time 1 hrs 20 mins
Total Assay Time 3 hrs 10 mins
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.