Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SCF/c-kit Ligand Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AB255NA
This item is not returnable.
View return policy
Description
SCF/c-kit Ligand Polyclonal specifically detects SCF/c-kit Ligand in Human samples. It is validated for Western Blot, Neutralization.Specifications
| SCF/c-kit Ligand | |
| Polyclonal | |
| Unconjugated | |
| P21583 | |
| KITLG | |
| E. coli-derived recombinant human SCF/c-kit Glu26-Ala189 Accession # P21583 | |
| 1 mg | |
| Primary | |
| Detects human SCF/c-kit Ligand in direct ELISAs and Western blots. In direct ELISA, approximately 45% cross-reactivity with recombinant mouse SCF is observed, approximately 30% cross-reactivity with recombinant canine SCF, recombinant rat SCF, and recombinant feline SCF is observed. It will also neutralize the biological activity of recombinant mouse SCF at a 10 fold higher IgG concentration. | |
| Human | |
| Purified |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 1 ug/mL, Neutralization 0.4-2 ug/mL | |
| c-kit Ligand, DCUA, DFNA69, DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, FPHH, KIT ligand, Kitl, KITLG, KL-1, Mast cell growth factor, MGF, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, SHEP7, SLF, steel factor | |
| Goat | |
| Protein A or G purified | |
| RUO | |
| 4254 | |
| Reconstitute at 1 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction