Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SCGF/CLEC11a Antibody, R&D Systems™
Mouse Monoclonal Antibody
$200.50
Specifications
Antigen | SCGF/CLEC11a |
---|---|
Clone | 239004 |
Dilution | Immunohistochemistry 5-25 ug/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB19041SP
|
R&D Systems
MAB19041SP |
25 μg |
Each of 1 for $200.50
|
|
|||||
Description
SCGF/CLEC11a Monoclonal antibody specifically detects SCGF/CLEC11a in Human Samples. It is validated for ImmunohistochemistrySpecifications
SCGF/CLEC11a | |
Immunohistochemistry 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
CLEC11a, CLECSF3, LSLCL, P47, SCGF | |
E. coli-derived recombiHumant human SCGF/CLEC11a, Gly19-Phe245, Accession # BAA21499 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
239004 | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
BAA21499 | |
6320 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title