Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SCGF/CLEC11a Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1904
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1904 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1904 Supplier R&D Systems Supplier No. MAB1904
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SCGF/CLEC11a Monoclonal specifically detects SCGF/CLEC11a in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.

Specifications

Antigen SCGF/CLEC11a
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 239029
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. BAA21499
Gene Alias CLEC11a, CLECSF3, LSLCL, P47, SCGF
Gene Symbols CLEC11A
Host Species Mouse
Immunogen E. coli-derived recombinant human SCGF/CLEC11a Gly19-Phe245 Accession # BAA21499
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6320
Test Specificity Detects human SCGF/CLEC11a in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant mouse SCGF-beta is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.