Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SCGF/CLEC11a Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems BAF1904
This item is not returnable.
View return policy
Description
SCGF/CLEC11a Polyclonal specifically detects SCGF/CLEC11a in Human samples. It is validated for Western Blot.Specifications
SCGF/CLEC11a | |
Polyclonal | |
Biotin | |
BAA21499 | |
CLEC11A | |
recombinant human SCGF-beta Gly19-Phe245 Accession # BAA21499 | |
50 μg | |
Primary | |
Detects human SCGF/CLEC11a in Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
CLEC11a, CLECSF3, LSLCL, P47, SCGF | |
Sheep | |
Affinity Purified | |
RUO | |
6320 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction