Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SCL/Tal1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3360
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3360 100 μg
MAB3360SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB3360 Supplier R&D Systems Supplier No. MAB3360
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SCL/Tal1 Monoclonal specifically detects SCL/Tal1 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen SCL/Tal1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 333803
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P17542
Gene Alias BHLHA17, bHLHa17tal-1, Class A basic helix-loop-helix protein 17, SCLT-cell acute lymphocytic leukemia protein 1, Stem cell protein, Tal1, TAL-1, T-cell acute lymphocytic leukemia 1, T-cell leukemia/lymphoma protein 5, TCL5, TCL5tal-1 product
Gene Symbols TAL1
Host Species Mouse
Immunogen E. coli-derived recombinant human SCL/Tal1 Arg199-Gly266 Accession # P17542
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6886
Test Specificity Detects human SCL/Tal1 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.