Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human SCP2 (aa 43-143) Control Fragment Recombinant Protein

Catalog No. RP94062
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μL

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55519 (PA5-55519. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes two proteins: sterol carrier protein X (SCPx) and sterol carrier protein 2 (SCP2), as a result of transcription initiation from 2 independently regulated promoters. The transcript initiated from the proximal promoter encodes the longer SCPx protein, and the transcript initiated from the distal promoter encodes the shorter SCP2 protein, with the 2 proteins sharing a common C-terminus. Evidence suggests that the SCPx protein is a peroxisome-associated thiolase that is involved in the oxidation of branched chain fatty acids, while the SCP2 protein is thought to be an intracellular lipid transfer protein. This gene is highly expressed in organs involved in lipid metabolism, and may play a role in Zellweger syndrome, in which cells are deficient in peroxisomes and have impaired bile acid synthesis. Alternative splicing of this gene produces multiple transcript variants, some encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P22307
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6342
Name Human SCP2 (aa 43-143) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AA409774; AA409893; C76618; C79031; NLTP; nonspecific lipid transfer protein; Non-specific lipid-transfer protein; NSLIPTR; ns-LTP; NSL-TP; propanoyl-CoA C-acyltransferase; SCP2; SCP-2; SCP-CHI; SCPX; SCP-X; Sterol carrier protein 2; sterol carrier protein 2, liver; sterol carrier protein X
Common Name SCP2
Gene Symbol SCP2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GKKALADAQIPYSAVDQACVGYVFGDSTCGQRAIYHSLGMTGIPIINVNNNCATGSTALFMARQLIQGGVAECVLALGFEKMSKGSLGIKFSDRTIPTDKH
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.