Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SCYL1 Partial ORF (AAH09967, 373 a.a. - 472 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7101531
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

This gene encodes a transcriptional regulator belonging to the SCY1-like family of kinase-like proteins. The protein has a divergent N-terminal kinase domain that is thought to be catalytically inactive, and can bind specific DNA sequences through its C-terminal domain. It activates transcription of the telomerase reverse transcriptase and DNA polymerase beta genes. The protein has been localized to the nucleus, and also to the cytoplasm and centrosomes during mitosis. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: DHKSSKSPESDWSSWEAEGSWEQGWQEPSSQEPPPDGTRLASEYNWGGPESSDKGDPFATLSARPSTQDRSRLSWPGRSARSGGGRWRPNAPRGRWPRAP

Specifications

Accession Number AAH09967
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 57410
Molecular Weight (g/mol) 36.63kDa
Name SCYL1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen DHKSSKSPESDWSSWEAEGSWEQGWQEPSSQEPPPDGTRLASEYNWGGPESSDKGDPFATLSARPSTQDRSRLSWPGRSARSGGGRWRPNAPRGRWPRAP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias GKLP/HT019/MGC78454/NKTL/NTKL/P105/TAPK/TEIF/TRAP
Common Name SCYL1
Gene Symbol SCYL1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.