Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SDHA Antibody, Novus Biologicals™
Mouse Monoclonal Antibody
$205.85 - $485.41
Specifications
Antigen | SDHA |
---|---|
Clone | 1084056 |
Dilution | Western Blot 1 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL |
Applications | Immunohistochemistry, Western Blot, Simple Western, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11586100
|
R&D Systems
MAB11586100 |
100 μg |
Each for $485.41
|
|
|||||
MAB11586SP
|
R&D Systems
MAB11586SP |
25 μg |
Each for $205.85
|
|
|||||
Description
SDHA Monoclonal antibody specifically detects SDHA in Human samples. It is validated for Immunohistochemistry,Western Blot,Simple Western,ImmunocytochemistrySpecifications
SDHA | |
Western Blot 1 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | |
CMD1GG, flavoprotein subunit of complex II, FP, MC2DN1, NDAXOA, PGL5, PPGL5, SDH1, SDH2, SDHF, succinate dehydrogenase complex, subunit A, flavoprotein (Fp) | |
Synthetic Peptide, Accession # P31040 | |
Primary | |
Reconstitute lyophilized material at 0.2 mg/ml in sterile PBS. For liquid material, refer to CoA for concentration. |
1084056 | |
Immunohistochemistry, Western Blot, Simple Western, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
P31040 | |
6389 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title