Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SEC13 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF9055100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF9055100 100 μg
AF9055SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF9055100 Supplier R&D Systems Supplier No. AF9055100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SEC13 Polyclonal specifically detects SEC13 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.

Specifications

Antigen SEC13
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.05 ug/mL, Simple Western 0.5 ug/mL, Immunocytochemistry 1-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P55735
Gene Alias D3S1231E, D3S1231ESEC13L1npp-20, Npp-20, protein SEC13 homolog, SEC13 (S. cerevisiae)-like 1, SEC13 homolog (S. cerevisiae), SEC13L1, SEC13-like 1 isoform, SEC13-like protein 1, SEC13R, SEC13-related protein, SEC13RSEC13-like 1 (S. cerevisiae)
Host Species Rabbit
Immunogen E. coli-derived recombinant human SEC13 Gly123-Gln322 Accession # P55735
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6396
Test Specificity Detects human SEC13 in direct ELISA and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.