Learn More
Human SEC13 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | SEC13 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.05 ug/mL, Simple Western 0.5 ug/mL, Immunocytochemistry 1-15 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. |
| Gene Accession No. | P55735 |
| Gene Alias | D3S1231E, D3S1231ESEC13L1npp-20, Npp-20, protein SEC13 homolog, SEC13 (S. cerevisiae)-like 1, SEC13 homolog (S. cerevisiae), SEC13L1, SEC13-like 1 isoform, SEC13-like protein 1, SEC13R, SEC13-related protein, SEC13RSEC13-like 1 (S. cerevisiae) |
| Host Species | Rabbit |
| Immunogen | E. coli-derived recombinant human SEC13 Gly123-Gln322 Accession # P55735 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.