Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SEC31A Full-length ORF (AAH47883.1, 1 a.a. - 509 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7099281
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is similar to yeast Sec31 protein. Yeast Sec31 protein is known to be a component of the COPII protein complex which is responsible for vesicle budding from endoplasmic reticulum (ER). This protein was found to colocalize with SEC13, one of the other components of COPII , in the subcellular structures corresponding to the vesicle transport function. An immunodepletion experiment confirmed that this protein is required for ER-Golgi transport. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: MKLKEVDRTAMQAWSPAQNHPIYLATGTSAQQLDATFSTNASLEIFELDLSDPSLDMKSCATFSSSHRYHKLIWGPYKMDSKGDVSGVLIAGGENGNIILYDPSKIIAGDKEVVIAQNDKHTGPVRALDVNIFQTNLVASGANESEIYIWDLNNFATPMTPGAKTQPPEDISCIAWNRQVQHILASASPSGRATVWDLRKNEPIIKVSDHSNRMHCSGLAWHPDVATQMVLASEDDRLPVIQMWDLRFASSPLRVLENHARGILAIAWSMADPELLLSCGKDAKILCSNPNTGEVLYELPTNTQWCFDIQWCPRNPAVLSAASFDGRISVYSIMGGSTDGLRQKQVDKLSSSFGNLDPFGTGQPLPPLQIPQQTAQHSIVLPLKKPPKWIRRPVGASFSFGGKLVTFENVRMPSHQGAEQQQQQHHVFISQVVTEKEFLSRSDQLQQAVQSQGFINYCQKKIDASQTEFEKNVWSFLKVNFEDDSRGKYLELLGYRKEDLGKKVNFWES

Specifications

Accession Number AAH47883.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 22872
Molecular Weight (g/mol) 83.1kDa
Name SEC31A (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MKLKEVDRTAMQAWSPAQNHPIYLATGTSAQQLDATFSTNASLEIFELDLSDPSLDMKSCATFSSSHRYHKLIWGPYKMDSKGDVSGVLIAGGENGNIILYDPSKIIAGDKEVVIAQNDKHTGPVRALDVNIFQTNLVASGANESEIYIWDLNNFATPMTPGAKTQPPEDISCIAWNRQVQHILASASPSGRATVWDLRKNEPIIKVSDHSNRMHCSGLAWHPDVATQMVLASEDDRLPVIQMWDLRFASSPLRVLENHARGILAIAWSMADPELLLSCGKDAKILCSNPNTGEVLYELPTNTQWCFDIQWCPRNPAVLSAASFDGRISVYSIMGGSTDGLRQKQVDKLSSSFGNLDPFGTGQPLPPLQIPQQTAQHSIVLPLKKPPKWIRRPVGASFSFGGKLVTFENVRMPSHQGAEQQQQQHHVFISQVVTEKEFLSRSDQLQQAVQSQGFINYCQKKIDASQTEFEKNVWSFLKVNFEDDSRGKYLELLGYRKEDLGKKVNFWES
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ABP125/ABP130/DKFZp686N07171/HSPC275/HSPC334/KIAA0905/MGC90305/SEC31L1
Common Name SEC31A
Gene Symbol SEC31A
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.