Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SECISBP2 Partial ORF (NP_076982.3, 106 a.a. - 205 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7101971
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

The incorporation of selenocysteine into a protein requires the concerted action of an mRNA element called a sec insertion sequence (SECIS), a selenocysteine-specific translation elongation factor and a SECIS binding protein. With these elements in place, a UGA codon can be decoded as selenocysteine. The gene described in this record encodes a nuclear protein that functions as a SECIS binding protein. Mutations in this gene have been associated with a reduction in activity of a specific thyroxine deiodinase, a selenocysteine-containing enzyme, and abnormal thyroid hormone metabolism. [provided by RefSeq]

Sequence: VPGSQYLYNQPSCYRGFQTVKHRNENTCPLPQEMKALFKKKTYDEKKTYDQQKFDSERADGTISSEIKSARGSHHLSIYAENSLKSDGYHKRTDRKSRII

Specifications

Accession Number NP_076982.3
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 79048
Molecular Weight (g/mol) 36.74kDa
Name SECISBP2 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen VPGSQYLYNQPSCYRGFQTVKHRNENTCPLPQEMKALFKKKTYDEKKTYDQQKFDSERADGTISSEIKSARGSHHLSIYAENSLKSDGYHKRTDRKSRII
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp686C09169/SBP2
Common Name SECISBP2
Gene Symbol SECISBP2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.