Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Semaphorin 4A Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB4694U100UG
This item is not returnable.
View return policy
Description
Semaphorin 4A Monoclonal specifically detects Semaphorin 4A in Human samples. It is validated for Flow Cytometry.Specifications
Semaphorin 4A | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9H3S1 | |
SEMA4A | |
Mouse myeloma cell line NS0-derived recombinant human Semaphorin 4A Gly32-His683 Accession # Q9H3S1 | |
RUO | |
64218 | |
Human | |
IgG1 |
Flow Cytometry | |
741531 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CORD10, RP35, Sema B, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4A, SEMA4A, SEMAB, semaphorin-4A, Semaphorin-B, SemB, SEMBFLJ12287 | |
Mouse | |
100 μg | |
Primary | |
Detects human Semaphorin 4A in direct ELISAs. In direct ELISAs, 50%-100% cross-reactivity with recombinant human (rh) Semaphorin 4C and rhSemaphorin 4G is observed, and less than 5% cross-reactivity with rhSemaphorin 4B, 4D, and recombinant mouse Semaphorin 4A is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction