Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Semaphorin 4C Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6125SP
This item is not returnable.
View return policy
Description
Semaphorin 4C Monoclonal specifically detects Semaphorin 4C in Human samples. It is validated for Western Blot.Specifications
| Semaphorin 4C | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q9C0C4 | |
| SEMA4C | |
| Mouse myeloma cell line NS0-derived recombinant human Semaphorin 4C Ala21-Gly663 (predicted) Accession # Q9C0C4 | |
| 25 μg | |
| Primary | |
| Detects human Semaphorin 4C in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Semaphorin 3A, 3B, 3C, 3D, 3E, 3F, 4A, 4B, 4D, 4G, 5A, 5B, 6A, 6B, 6C, 6D, 7A, recombinant mouse Semaphorin 3B, 3C, 3E, 3F, 4A, 4B, 4C, 4D, 4F, 4G, 6A, 6B, 6C, 6D, or 7A is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 660016 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ20369, KIAA1739, MGC126382, MGC126383, M-Sema F, M-SEMA-F, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4C, SEMA4C, SEMACL1, SEMAF, SEMAI, semaphorin-4C | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 54910 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction