Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Semaphorin 4D/CD100 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB74701R100UG
This item is not returnable.
View return policy
Description
Semaphorin 4D/CD100 Monoclonal specifically detects Semaphorin 4D/CD100 in Human samples. It is validated for Flow Cytometry.Specifications
Semaphorin 4D/CD100 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q92854 | |
SEMA4D | |
Chinese hamster ovary cell line CHO-derived recombinant human Semaphorin 4D/CD100 Met22-Arg734 Accession # Q92854 | |
RUO | |
10507 | |
Human | |
IgG1 |
Flow Cytometry | |
758726 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
A8, BB18, CD100, CD100 antigen, CD100M-sema-G, coll-4, FLJ33485, FLJ34282, FLJ39737, FLJ46484, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4D, MGC169138, MGC169141, M-sema-G, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4D, SEMA4D, SEMAJ, semaphorin-4D | |
Mouse | |
100 μg | |
Primary | |
Detects human Semaphorin 4D/CD100 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Semaphorin 4B, 4C, 4G, or recombinant mouse Semaphorin 4D is observed. In Western blots, no cross-reactivity with rhSemaphorin 4A, 6A, or 6C is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction