Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Semaphorin 4G Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF5840
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF5840 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF5840 Supplier R&D Systems Supplier No. BAF5840
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Semaphorin 4G Polyclonal specifically detects Semaphorin 4G in Human samples. It is validated for Western Blot.

Specifications

Antigen Semaphorin 4G
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q9NTN9
Gene Alias KIAA1619FLJ20590, MGC102867, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4G, SEMA4G, semaphorin-4G
Gene Symbols SEMA4G
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Semaphorin 4G Val18-Leu675 Accession # Q9NTN9
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57715
Test Specificity Detects human Semaphorin 4G in Western blots. In Western blots, approximately 50% cross-reactivity with recombinant mouse Semaphorin 4G is observed and less than 1% cross reactivity with recombinant human (rh) Semaphorin 3B, rhSemaphorin 4A, and rhSemaphorin 4C is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.