Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Semaphorin 6A Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
$200.50 - $549.50
Specifications
| Antigen | Semaphorin 6A |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF |
| Classification | Polyclonal |
Description
Semaphorin 6A Polyclonal specifically detects Semaphorin 6A in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
| Semaphorin 6A | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready | |
| Polyclonal | |
| Goat | |
| Human | |
| KIAA1368HT018, SEMA, sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A, Sema VIA, Sema6A, SEMA6A1, SEMA6A-1, semaphorin 6A-1, Semaphorin VIA, semaphorin-6A, Semaphorin-6A-1, SEMAQ, VIA | |
| SEMA6A | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
| Unconjugated | |
| RUO | |
| Q9H2E6 | |
| 57556 | |
| Mouse myeloma cell line NS0-derived recombinant human Semaphorin 6A Gly19-Thr649 Accession # Q9H2E6 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Semaphorin 6A in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant mouse Semaphorin 6A is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title