Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Semaphorin 6C Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2219
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2219 100 μg
AF2219SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2219 Supplier R&D Systems Supplier No. AF2219
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Semaphorin 6C Polyclonal specifically detects Semaphorin 6C in Human samples. It is validated for Western Blot.

Specifications

Antigen Semaphorin 6C
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q9H3T2
Gene Alias KIAA1869, m-SemaY, m-SemaY2, sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6C, Sema Y, Sema6C, Semaphorin Y, semaphorin-6C, semaphorin-Y, SEMAY
Gene Symbols SEMA6C
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Semaphorin 6C Ala25-Val601 Accession # Q9H3T2
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10500
Test Specificity Detects human Semaphorin 6C in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) Sema 3A, rhSema 3B, rhSema 6A, rhSema 6B, rhSema 7A, recombinant mouse (rm) Sema 3C, rmSema 3E, and rmSema 3F is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.