Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Semaphorin 6D Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2095SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2095SP 25 μg
AF2095 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2095SP Supplier R&D Systems Supplier No. AF2095SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Semaphorin 6D Polyclonal specifically detects Semaphorin 6D in Human samples. It is validated for Western Blot.

Specifications

Antigen Semaphorin 6D
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8NFY4
Gene Alias KIAA1479FLJ11598, sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6D, Sema6D, semaphorin-6D
Gene Symbols SEMA6D
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Semaphorin 6D Ser22-His660 Accession # Q8NFY4
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80031
Test Specificity Detects human Semaphorin 6D in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) Semaphorin 6C is observed, 5% cross-reactivity with rhSemaphorin 6B is observed and less than 1% cross-reactivity with rhSemaphorin 3A, rhSemaphorin 3B, recombinant mouse (rm) Semaphorin 3E, rmSemaphorin 3F, rhSemaphorin 3A, and rhSemaphorin 7A is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.