Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Semaphorin 7A Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
$200.50 - $549.50
Specifications
| Antigen | Semaphorin 7A |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, CyTOF |
| Classification | Polyclonal |
Description
Semaphorin 7A Polyclonal specifically detects Semaphorin 7A in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
| Semaphorin 7A | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Polyclonal | |
| Goat | |
| Human | |
| CD108, CD108 antigen, CD108MGC126696, CDw108, H-SEMA-K1, H-Sema-L, JMH, JMH blood group antigen, John-Milton-Hargen human blood group Ag, MGC126692, sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin)7A, sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin)7A (JMH blood group), sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, 7A, Sema K1, Sema L, Sema7A, SEMAK1, SEMAL, SEMALJohn Milton Hagen blood group H-Sema K1, semaphorin 7A, GPI membrane anchor (John Milton Hagen blood group), semaphorin K1, semaphorin L, semaphorin-7A, semaphorin-K1, Semaphorin-L | |
| SEMA7A | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Flow Cytometry, CyTOF | |
| Unconjugated | |
| RUO | |
| O75326 | |
| 8482 | |
| Mouse myeloma cell line NS0-derived recombinant human Semaphorin 7A His47-Ala648 Accession # O75326 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Semaphorin 7A in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse Semaphorin 7A is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title