Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Semaphorin 7A Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF2068
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF2068 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF2068 Supplier R&D Systems Supplier No. BAF2068
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Semaphorin 7A Polyclonal specifically detects Semaphorin 7A in Human samples. It is validated for Western Blot.

Specifications

Antigen Semaphorin 7A
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. O75326
Gene Alias CD108, CD108 antigen, CD108MGC126696, CDw108, H-SEMA-K1, H-Sema-L, JMH, JMH blood group antigen, John-Milton-Hargen human blood group Ag, MGC126692, sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin)7A, sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin)7A (JMH blood group), sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, 7A, Sema K1, Sema L, Sema7A, SEMAK1, SEMAL, SEMALJohn Milton Hagen blood group H-Sema K1, semaphorin 7A, GPI membrane anchor (John Milton Hagen blood group), semaphorin K1, semaphorin L, semaphorin-7A, semaphorin-K1, Semaphorin-L
Gene Symbols SEMA7A
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Semaphorin 7A His47-Ala648 Accession # O75326
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8482
Test Specificity Detects human Semaphorin 7A in Western blots. In Western blots, approximately 50% cross-reactivity with recombinant mouse (rm) Semaphorin 7A is observed and less than 1% cross-reactivity with recombinant human Semaphorin 3A, 6B, 6D and rmSema 3C, 3E, and 3F is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.