Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Semaphorin-7A ELISA Kit

Catalog No. p-7219619
Encompass_Preferred
Click to view available options
Quantity:
5 x 96 Tests
96 Tests

ELISA

Human Semaphorin-7A quantitates human Semaphorin-7A in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Semaphorin-7A.

CD108 (Sema7A) is a GPI-anchored semaphorin family member, which enhances central and peripheral axonal growth and is required for proper axon track formation during ebryogenesis. CD108 also regulates osteoclast differentiation and pre-osteoblastic cell migration, and in immune system affects cell proliferation, chemotaxis and cytokine release. On erythrocytes CD108 defines the JMH (John-Milton-Hagen) human blood group. CD108 signalizes through its receptors - plexin C1 and beta1 integrins.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 2900057C09Rik; Cd108; CDw108; H-Sema K1; H-SEMA-K1; H-Sema-L; JMH; JMH blood group antigen; John Milton Hagen blood group H-Sema K1; john-Milton-Hargen human blood group Ag; MGC126692; MGC126696; M-Sema-L; sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin) 7 A; sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin) 7 A (JMH blood group); sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, 7 A; sema K1; Sema L; Sema7a; SEMAK1; Semal; semaphorin 7 A (John Milton Hagen blood group); semaphorin 7 A, GPI membrane anchor; semaphorin 7 A, GPI membrane anchor (John Milton Hagen blood group); Semaphorin K1; semaphorin-7 A; Semaphorin-K1; semaphorin-L; Semk1
Assay Range 0.16 to 40 ng/mL
Assay Sensitivity 0.16 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 8482
Gene Symbol SEMA7A
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target SEMA7A
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.