Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Serpin A1/alpha 1-Antitrypsin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1268SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1268SP 25 μg
MAB1268100 100 μg
MAB1268500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB1268SP Supplier R&D Systems Supplier No. MAB1268SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 3 publications

Serpin A1/alpha 1-Antitrypsin Monoclonal specifically detects Serpin A1/alpha 1-Antitrypsin in Human samples. It is validated for Western Blot, Simple Western, Immunoprecipitation, Immunocytochemistry.

Specifications

Antigen Serpin A1/alpha 1-Antitrypsin
Applications Western Blot, Immunoprecipitation, Immunocytochemistry
Classification Monoclonal
Clone 202808
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P01009
Gene Alias A1A, A1AT, AATMGC23330, alpha 1-Antitrypsin, alpha 1-Proteinase Inhibitor, Alpha-1 protease inhibitor, Alpha-1-antiproteinase, alpha-1-antitrypsin, antitrypsin), member 1, member 1, PIMGC9222, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 1, Serpin A1, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin)
Gene Symbols SERPINA1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Serpin A1/alpha 1-Antitrypsin Glu25-Lys418 Accession # P01009
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5265
Test Specificity Detects human Serpin A1/alpha 1-Antitrypsin in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Serpin A3, A4, A5, C1, F1, recombinant mouse Serpin D1, or F2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.