Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Serpin A1/alpha 1-Antitrypsin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF12681SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF12681SP 25 μg
AF12681100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF12681SP Supplier R&D Systems Supplier No. AF12681SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Antibody

Serpin A1/alpha 1-Antitrypsin specifically detects Serpin A1/alpha 1-Antitrypsin in Human samples. It is validated for Western Blot.

Specifications

Antigen SerpinA1/alpha1-Antitrypsin
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.1μg/mL
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Gene Accession No. P01009
Gene Alias A1A, A1AT, AATMGC23330, alpha 1-Antitrypsin, alpha 1-Proteinase Inhibitor, Alpha-1 protease inhibitor, Alpha-1-antiproteinase, alpha-1-antitrypsin, antitrypsin), member 1, member 1, PIMGC9222, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 1, Serpin A1, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin)
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Serpin A1/alpha 1-Antitrypsin Glu25-Lys418 Accession # P01009
Purification Method Affinity Purified
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 5265
Test Specificity Detects human Serpin A1/alpha 1-Antitrypsin in direct ELISAs.
Reconstitution Reconstitute at 0.2mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.