Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Serpin A1/alpha 1-Antitrypsin Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $444.50
Specifications
| Antigen | Serpin A1/alpha 1-Antitrypsin |
|---|---|
| Clone | 1019625 |
| Dilution | Western Blot 2 ug/mL, Simple Western 20 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB12681SP
![]() |
R&D Systems
MAB12681SP |
25 μg |
Each for $200.50
|
|
|||||
MAB12681100
![]() |
R&D Systems
MAB12681100 |
100 μg |
Each for $444.50
|
|
|||||
Description
Serpin A1/alpha 1-Antitrypsin Monoclonal specifically detects Serpin A1/alpha 1-Antitrypsin in Human samples. It is validated for Western Blot, Simple Western.Specifications
| Serpin A1/alpha 1-Antitrypsin | |
| Western Blot 2 ug/mL, Simple Western 20 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| A1A, A1AT, AATMGC23330, alpha 1-Antitrypsin, alpha 1-Proteinase Inhibitor, Alpha-1 protease inhibitor, Alpha-1-antiproteinase, alpha-1-antitrypsin, antitrypsin), member 1, member 1, PIMGC9222, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 1, Serpin A1, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin) | |
| Mouse myeloma cell line, NS0-derived human Serpin A1/alpha 1-Antitrypsin, Glu25-Lys418, Accession # P01009 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1019625 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P01009 | |
| 5265 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title