Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Serpin A1/alpha 1-Antitrypsin ELISA Kit (Colorimetric)
SDP

Catalog No. NB397847 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
96 wells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397847 96 wells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397847 Supplier Novus Biologicals™ Supplier No. NBP312165
Add to Cart
Add to Cart

Kit (Colorimetric)

Specifications

Assay Range 0.24 to 200 ng/mL
Immunoassay Kit Format Colorimetric Detection Kit
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Colostrum, Milk
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 5265
Gene Symbol SERPINA1
Quantity 96 wells
Research Discipline Stem Cell Markers
Storage Requirements Store at 4°C.
Synonym A1A, AATMGC23330, Alpha-1 protease inhibitor, Alpha-1-antiproteinase, alpha-1-antitrypsin, antitrypsin), member 1, member 1, PIMGC9222, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 1, Serpin A1, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin)
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.