Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Serpin A10/ZPI Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $549.50
Specifications
Antigen | Serpin A10/ZPI |
---|---|
Clone | 895910 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB8115SP
![]() |
R&D Systems
MAB8115SP |
25 μg |
Each for $200.50
|
|
|||||
MAB8115100
![]() |
R&D Systems
MAB8115100 |
100 μg |
Each for $549.50
|
|
|||||
Description
Serpin A10/ZPI Monoclonal specifically detects Serpin A10/ZPI in Human samples. It is validated for Western Blot.Specifications
Serpin A10/ZPI | |
Western Blot 1 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
Q9UK55 | |
51156 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
895910 | |
Western Blot | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
antitrypsin), member 10, member 10, PZI, PZIprotein Z-dependent protease inhibitor, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A10, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), ZPI, ZPIPZ-dependent protease inhibitor | |
Chinese hamster ovary cell line CHO-derived recombinant human Serpin-A10/ZPI Met1-Leu444 Accession # Q9UK55 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Serpin A10/ZPI in direct ELISAs and Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title