Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Serpin A12 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4410SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4410SP 25 μg
MAB4410 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB4410SP Supplier R&D Systems Supplier No. MAB4410SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Serpin A12 Monoclonal specifically detects Serpin A12 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Serpin A12
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 484223
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8IW75
Gene Alias antitrypsin), member 12, member 12, OL-64, OL-64Visceral adipose-specific serpin, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serpin A12, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Vaspin, Visceral adipose tissue-derived serine protease inhibitor
Gene Symbols SERPINA12
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Serpin A12 Leu20-Lys414 Accession # Q8IW75
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145264
Test Specificity Detects human Serpin A12 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) Serpin A1, A3, A4, A5, A6, B8, 9, C1, D1, E1, F2, G1, I1, I2, recombinant mouse (rm) Serpin E2, F1, recombinant rat Serpin B6, rhA1 Antitrypsin, rhAngiotensinogin, or rmPEDF is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.