Learn More
Description
Includes
Human Serpin A12 microplate, 96-well; Human Serpin A12 conjugate; Human Serpin A12 standard; Buffers and diluents, Color reagents A and B; Stop solution; Plate sealers
Specifications
Specifications
| Product Type | ELISA Kit |
| Quantity | 1 Kit |
| Species | Human |
| Conjugate | HRP |
| Assay Sensitivity | 14.6 pg/mL |
| Assay Range | 31.30 to 2000 pg/mL |
| For Use With (Application) | ELISA |
| Product Line | R&D Systems |
| Sample Type | Cell Culture Supernates (50uL), Serum (50uL), EDTA Plasma (50uL), Heparin Plasma (50uL), Saliva (50uL), Urine (50uL), Human Milk (50uL) |
| Synonym | antitrypsin), member 12, member 12, OL-64, OL-64Visceral adipose-specific serpin, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serpin A12, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Vaspin, Visceral adipose tissue-derived serine protease inhibitor |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
