Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Serpin A3/alpha 1-Antichymotrypsin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1295SP
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1295SP 25 μg
MAB1295 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1295SP Supplier R&D Systems Supplier No. MAB1295SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Serpin A3/alpha 1-Antichymotrypsin Monoclonal specifically detects Serpin A3/alpha 1-Antichymotrypsin in Human samples. It is validated for Western Blot.

Specifications

Antigen Serpin A3/alpha 1-Antichymotrypsin
Applications Western Blot
Classification Monoclonal
Clone 213907
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P01011
Gene Alias AACTMGC88254, ACTGIG25, alpha 1-Antichymotrypsin, alpha-1-antichymotrypsin, antitrypsin), member 3, Cell growth-inhibiting gene 24/25 protein, growth-inhibiting protein 24, growth-inhibiting protein 25, member 3, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 3, Serpin A3, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin)
Gene Symbols SERPINA3
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Serpin A3 aa 26-423 Accession # P01011
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12
Test Specificity Detects recombinant human (rh)Serpin A3 in direct ELISAs and Western blots. In these formats, this antibody shows 5 - 25% cross-reactivity with rhSerpins A1 and A5 but no cross-reactivity with rhSerpins C1, D1, or F2.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.