Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Serpin A3/alpha 1-Antichymotrypsin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1295
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1295 100 μg
AF1295SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF1295 Supplier R&D Systems Supplier No. AF1295
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Serpin A3/alpha 1-Antichymotrypsin Polyclonal specifically detects Serpin A3/alpha 1-Antichymotrypsin in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Serpin A3/alpha 1-Antichymotrypsin
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Gene Accession No. P01011
Gene Alias AACTMGC88254, ACTGIG25, alpha 1-Antichymotrypsin, alpha-1-antichymotrypsin, antitrypsin), member 3, Cell growth-inhibiting gene 24/25 protein, growth-inhibiting protein 24, growth-inhibiting protein 25, member 3, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 3, Serpin A3, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin)
Gene Symbols SERPINA3
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Serpin A3/alpha 1-Antichymotrypsin Asn26-Ala423 Accession # P01011
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12
Test Specificity Detects human Serpin A3/alpha 1-Antichymotrypsin in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse Serpin A1a is observed and less than 3% cross-reactivity with recombinant human (rh) Serpin A1, rhSerpin A4, rhSerpin A5 and rhSerpin A9 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.