Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Serpin A5/Protein C Inhibitor Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1266SP
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1266SP 25 μg
MAB1266 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1266SP Supplier R&D Systems Supplier No. MAB1266SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

Serpin A5/Protein C Inhibitor Monoclonal specifically detects Serpin A5/Protein C Inhibitor in Human samples. It is validated for Western Blot, Immunoprecipitation, Intracellular Staining by Flow Cytometry, CyTOF-ready.

Specifications

Antigen Serpin A5/Protein C Inhibitor
Applications Western Blot, Immunoprecipitation, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 210326
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P05154
Gene Alias Acrosomal serine protease inhibitor, antitrypsin), member 5, member 5, PAI-3, PAI3PLANH3, PCI, PCIplasminogen activator inhibitor III, Plasminogen activator inhibitor 3, plasminogen activator inhibitor-3, PROCIplasma serine protease inhibitor, Protein C inhibitor, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A5, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), SERPNA5
Gene Symbols SERPINA5
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Serpin A5/Protein C His20-Pro406 Accession # P05154
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5104
Test Specificity Detects human Serpin A5/Protein C in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Serpin A1, A3, F2, C1, or recombinant mouse Serpin A5 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.