Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Serpin A8/Angiotensinogen Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3156
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3156 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3156 Supplier R&D Systems Supplier No. AF3156
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 3 publications

Serpin A8/Angiotensinogen Polyclonal specifically detects Serpin A8/Angiotensinogen in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.

Specifications

Antigen Serpin A8/Angiotensinogen
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 2-15 ug/mL
Gene Accession No. ABM85122
Gene Alias AGT, alpha-1 antiproteinase, antitrypsin, angiotensin II, Angiotensinogen, angiotensinogen (serpin peptidase inhibitor, clade A, member 8), ANHU, Aogen, FLJ92595, FLJ97926, PAT, pre-angiotensinogen, serine (or cysteine) proteinase inhibitor, Serpin A8, SERPINA8angiotensin I
Gene Symbols AGT
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Serpin A8/Angiotensinogen Asp34-Ala485 Accession # ABM85122
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 183
Test Specificity Detects human Serpin A8/Angiotensinogen in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse Serpin A8/Angiotensinogen is observed, and less than 5% cross-reactivity with recombinant human Serpin A1, A4, A5, F2, G1, I1, and I2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.