Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Serpin A9/Centerin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5429
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5429 100 μg
MAB5429SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB5429 Supplier R&D Systems Supplier No. MAB5429
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Serpin A9/Centerin Monoclonal specifically detects Serpin A9/Centerin in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Serpin A9/Centerin
Applications Immunohistochemistry
Classification Monoclonal
Clone 585302
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAQ89063
Gene Alias Centerin, GCET1, SERPINA11, SERPINA11b
Gene Symbols SERPINA9
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Serpin A9/Centerin Ala24-Thr417 Accession # AAQ89063
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 327657
Test Specificity Detects human Serpin A9/Centerin in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Serpin A1, A3, A4, A5, A6, A11, A12, B6, B8, B9, C1, D1, E1, E2, F1, F2, G1, I1, I2, recombinant mouse (rm) SerpinA3N, and rmSerpinA9 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.