Learn More
Human Serpin B2 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | SerpinB2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 930109 |
| Conjugate | Alexa Fluor 488 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Gene Accession No. | P05120 |
| Gene Alias | HsT1201, Monocyte Arg-serpin, PAI, PAI2, PAI2Urokinase inhibitor, Placental plasminogen activator inhibitor, PLANH2, PLANH2PAI-2, plasminogen activator inhibitor 2, plasminogen activator inhibitor, type II (arginine-serpin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 2, Serpin B2, serpin peptidase inhibitor, clade B (ovalbumin), member 2, Urokinase Inhibitor |
| Host Species | Mouse |
| Show More |
For Research Use Only (RUO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.