Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Serpin B2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8550
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8550 100 μg
MAB8550SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB8550 Supplier R&D Systems Supplier No. MAB8550
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SerpinB2 Monoclonal specifically detects SerpinB2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready, Knockout Validated.

Specifications

Antigen SerpinB2
Applications Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF, KnockDown
Classification Monoclonal
Clone 930109
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready, Knockout Validated
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P05120
Gene Alias HsT1201, Monocyte Arg-serpin, PAI, PAI2, PAI2Urokinase inhibitor, Placental plasminogen activator inhibitor, PLANH2, PLANH2PAI-2, plasminogen activator inhibitor 2, plasminogen activator inhibitor, type II (arginine-serpin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 2, Serpin B2, serpin peptidase inhibitor, clade B (ovalbumin), member 2, Urokinase Inhibitor
Gene Symbols SERPINB2
Host Species Mouse
Immunogen E. coli-derived recombinant human Serpin B2 Glu2-Ala65, Ala99-Pro415 Accession # P05120
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5055
Test Specificity Detects human Serpin B2 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.