Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Serpin B2 Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Supplier: R&D Systems AF8550
This item is not returnable.
View return policy
Description
SerpinB2 Polyclonal specifically detects SerpinB2 in Human samples. It is validated for Western Blot, Simple Western, Knockout Validated.Specifications
SerpinB2 | |
Polyclonal | |
Western Blot 1:1000 dilution, Simple Western 1:100 dilution, Knockout Validated 2 ug/mL | |
P05120 | |
SERPINB2 | |
E. coli-derived recombinant human Serpin B2 Glu2-Pro415 (Asn66-Gln98 del) Accession # P05120 | |
100 μL | |
Primary | |
Detects human Serpin B2 in direct ELISAs and Western blots. In direct ELISAs, less than 3% cross-reactivity with recombinant human (rh) Serpin B6, rhSerpin B8, and rhSerpin B9 is observed. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. | |
IgG |
Western Blot, KnockDown | |
Unconjugated | |
Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. with Sodium Azide | |
HsT1201, Monocyte Arg-serpin, PAI, PAI2, PAI2Urokinase inhibitor, Placental plasminogen activator inhibitor, PLANH2, PLANH2PAI-2, plasminogen activator inhibitor 2, plasminogen activator inhibitor, type II (arginine-serpin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 2, Serpin B2, serpin peptidase inhibitor, clade B (ovalbumin), member 2, Urokinase Inhibitor | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
5055 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction