Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Serpin B5/Maspin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2218
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2218 100 μg
MAB2218SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2218 Supplier R&D Systems Supplier No. MAB2218
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Serpin B5/Maspin Monoclonal specifically detects Serpin B5/Maspin in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Serpin B5/Maspin
Applications Western Blot, Immunoprecipitation, Immunoassay
Classification Monoclonal
Clone 305339
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL, Knockout Validated
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P36952
Gene Alias maspin, Peptidase inhibitor 5, PI5, PI-5, PI5protease inhibitor 5 (maspin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 5, serpin B5, serpin peptidase inhibitor, clade B (ovalbumin), member 5
Gene Symbols SERPINB5
Host Species Rat
Immunogen E. coli-derived recombinant human Serpin B5 Met1-Pro375 Accession # P36952
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5268
Test Specificity Detects human Serpin B5/Maspin in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Serpin A5, B6, B8 or B9 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.