Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Serpin B5/Maspin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2218SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2218SP 25 μg
AF2218 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2218SP Supplier R&D Systems Supplier No. AF2218SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Serpin B5/Maspin Polyclonal specifically detects Serpin B5/Maspin in Human samples. It is validated for Western Blot, Knockout Validated.

Specifications

Antigen Serpin B5/Maspin
Applications Western Blot, Immunoassay
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Knockout Validated
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P36952
Gene Alias maspin, Peptidase inhibitor 5, PI5, PI-5, PI5protease inhibitor 5 (maspin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 5, serpin B5, serpin peptidase inhibitor, clade B (ovalbumin), member 5
Gene Symbols SERPINB5
Host Species Goat
Immunogen E. coli-derived recombinant human Serpin B5/Maspin Met1-Pro375 Accession # P36952
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5268
Test Specificity Detects human Serpin B5/Maspin in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) Serpin A1, rhSerpin A3, rhSerpin A4, rhSerpin A5, rhSerpin C1, rhSerpin F2, rhSerpin I1, rhSerpin I2, rhPEDF, and recombinant mouse Serpin D1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.