Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Serpin B6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4336
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4336 100 μg
AF4336SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF4336 Supplier R&D Systems Supplier No. AF4336
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Serpin B6 Polyclonal specifically detects Serpin B6 in Human samples. It is validated for Western Blot.

Specifications

Antigen Serpin B6
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. P35237
Gene Alias CAP, CAPMGC111370, Cytoplasmic antiproteinase, deafness, autosomal recessive 91, DFNB91, Peptidase inhibitor 6, PI6, PI-6, PI6MSTP057, Placental thrombin inhibitor, protease inhibitor 6 (placental thrombin inhibitor), PTI, PTIDKFZp686I04222, serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 6, serpin B6, serpin peptidase inhibitor, clade B (ovalbumin), member 6, SPI3
Gene Symbols SERPINB6
Host Species Sheep
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Serpin B6 Asp2-Pro376 Accession # P35237
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5269
Test Specificity Detects human Serpin B6 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) Serpin B8 and rhSerpin B9 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.